Hydroquinone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11411-0B
A11411-0I
A11411-30
CAAAAA11411-0BEA
124.5
CAD
CAAAAA11411-0B
CAAAAA11411-0I
CAAAAA11411-30
Hydroquinone 99%
Hydroquinone
Purity: 99%
CAS No.: 123-31-9
Molecular Formula: C6H6O2
Formula Weight: 110.11
Flash Point: 165°(329°F)
Odor: Odorless
pH: 3.75
Melting Point: 170-172°
Boiling Point: 285-287°
Beilstein Reference: 605970
Merck Reference: 14,4808
UN No.: 3077
MDL No.: MFCD00002339
Formula:
C₆H₄(OH)₂ Boiling Pt: 285…287 °C Melting Pt: 170…172 °C Density: 1.320 Flash Pt: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 14,4808 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...