Cyclohexene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11359-0F
A11359-AE
A11359-AP
CAAAAA11359-0FEA
254.38
CAD
CAAAAA11359-0F
CAAAAA11359-AE
CAAAAA11359-AP
Cyclohexene 99%
Cyclohexene
Purity: 99%
CAS No.: 110-83-8
Molecular Formula: C6H10
Formula Weight: 82.15
Flash Point: -20°(-4°F)
Refractive Index: 1.4460
Odor: Petrol-like
Melting Point: -104°
Boiling Point: 83-85°
Beilstein Reference: 906737
Merck Reference: 14,2727
UN No.: 2256
MDL No.: MFCD00001539
Formula:
C₆H₁₀ MW: 82.15 g/mol Boiling Pt: 83…85 °C Melting Pt: –104 °C Density: 0.811 Flash Pt: –20 °C (–4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001539 CAS Number: 110-83-8 EINECS: 203-807-8 UN: 2256 ADR: 3,II Merck Index: 14,2727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...