1-Butynylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11332-06
A11332-09
A11332-14
CAAAAA11332-06EA
58.14
CAD
CAAAAA11332-06
CAAAAA11332-09
CAAAAA11332-14
1-Butynylbenzene 98%
1-Butynylbenzene
Purity: 98%
CAS No.: 622-76-4
Molecular Formula: C10H10
Formula Weight: 130.19
Flash Point: 68°(154°F)
Refractive Index: 1.5520
Boiling Point: 201-203°
Beilstein Reference: 1903397
MDL No.: MFCD00039945
Formula:
C₆H₅CCCH₂CH₃ Boiling Pt: 201…203 °C Density: 0.918 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039945 CAS Number: 622-76-4 EINECS: 210-752-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...