Mesitylene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3,5-Trimethylbenzene
A11323-0F
A11323-AE
A11323-AP
CAAAAA11323-0FEA
553
CAD
CAAAAA11323-0F
CAAAAA11323-AE
CAAAAA11323-AP
Mesitylene 98+%
Mesitylene
Purity: 98+%
CAS No.: 108-67-8
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 44°(111°F)
Refractive Index: 1.4990
Odor: Unpleasant
Melting Point: -45°
Boiling Point: 163-165°
Beilstein Reference: 906806
Merck Reference: 14,5907
UN No.: 2325
MDL No.: MFCD00008538
Formula:
C₉H₁₂ MW: 120.19 g/mol Boiling Pt: 163…165 °C Melting Pt: –45 °C Density: 0.865 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008538 CAS Number: 108-67-8 EINECS: 203-604-4 UN: 2325 ADR: 3,III Merck Index: 14,5907 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...