1,6-Heptadiyne 97%

Supplier: Thermo Scientific Chemicals
Danger

A11318-06 A11318-14 A11318-22
CAAAAA11318-06EA 178.54 CAD
CAAAAA11318-06 CAAAAA11318-14 CAAAAA11318-22
1,6-Heptadiyne 97%
1,6-Heptadiyne

Purity: 97% CAS No.: 2396-63-6 Molecular Formula: C7H8 Formula Weight: 92.14 Storage Temperature: 2°C to 8°C Flash Point: 9°(48°F) Refractive Index: 1.4420 Odor: Recognizable Boiling Point: 109-111° Beilstein Reference: 1731756 UN No.: 3295 MDL No.: MFCD00014925

Formula: C₇H₈
MW: 92.14 g/mol
Boiling Pt: 109…111 °C
Melting Pt: –85 °C
Density: 0.810
Flash Pt: 9 °C (48 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00014925
CAS Number: 2396-63-6
EINECS: 219-253-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR