Paraformaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger

A11313-0C A11313-0E A11313-22 A11313-36
CAAA-A11313-0CEA 464.52 CAD
CAAA-A11313-0C CAAA-A11313-0E CAAA-A11313-22 CAAA-A11313-36
Paraformaldehyde 97%
Paraformaldehyde
Formula: (CH₂O)n
Boiling Pt: 135 - 136 °C (13 -rr)
Melting Pt: 120…170 °C (dec.)
Density: 1.30
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4
EINECS: 200-001-8
UN: 2213
ADR: 4.1,III
Merck Index: 14,7025

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR