Potassium periodate 99%

Supplier: Thermo Scientific Chemicals
Danger

A11308-0E A11308-22 A11308-36
CAAAAA11308-0EEA 1584.74 CAD
CAAAAA11308-0E CAAAAA11308-22 CAAAAA11308-36
Potassium periodate 99%
Potassium periodate

Purity: 99% CAS No.: 7790-21-8 Molecular Formula: KIO4 Formula Weight: 230.00 Melting Point: 582° Merck Reference: 14,7654 MDL No.: MFCD00011407

Formula: KlO₄
Melting Pt: 582 °C
Density: 3.618
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,III
Merck Index: 14,7654

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR