Barium nitrate 99%

Supplier: Thermo Scientific Chemicals
Danger

A11305-0E A11305-36
CAAAAA11305-0EEA 306.52 CAD
CAAAAA11305-0E CAAAAA11305-36
Barium nitrate 99%
Barium nitrate

Purity: 99% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Melting Point: 592° dec. Merck Reference: 14,983 MDL No.: MFCD00003442

Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: 592 °C (dec.)
Density: 3.24
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 14,983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR