α-Tropolone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone

A11299-03 A11299-06 A11299-14
CAAAAA11299-03EA 69.36 CAD
CAAAAA11299-03 CAAAAA11299-06 CAAAAA11299-14
α-Tropolone 98%
α-Tropolone

A non-benzenoid aromatic compound


Purity: 98% CAS No.: 533-75-5 Molecular Formula: C7H6O2 Formula Weight: 122.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 50-55° Boiling Point: 115°/12mm Beilstein Reference: 1904978 MDL No.: MFCD00004158

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 115 °C/12 mm
Melting Pt: 50…55 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR