Copper(II) sulfate pentahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11262-0B
A11262-0G
A11262-0I
CAAAAA11262-0BEA
121.5
CAD
CAAAAA11262-0B
CAAAAA11262-0G
CAAAAA11262-0I
Copper(II) sulfate pentahydrate 99%
Copper (II) sulphate pentahydrate
Purity: 99%
CAS No.: 7758-99-8
Molecular Formula: CuSO4.5H2O
Formula Weight: 249.68 (159.60anhy)
Odor: Odorless
pH: 4
Merck Reference: 14,2653
UN No.: 3288
MDL No.: MFCD00149681
Formula:
CuSO₄·5H₂O Density: 2.270 Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3077 ADR: 9,III REACH: 01-2119520566-40 Merck Index: 14,2653 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...