Sodium perchlorate monohydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11243-0B
A11243-0I
A11243-30
CAAAAA11243-0BEA
301.78
CAD
CAAAAA11243-0B
CAAAAA11243-0I
CAAAAA11243-30
Sodium perchlorate monohydrate 97+%
Sodium perchlorate monohydrate
Purity: 97+%
CAS No.: 7791-07-3
Molecular Formula: NaClO4.H2O
Formula Weight: 140.48 (122.46anhy)
Odor: Odorless
Melting Point: 130° dec.
UN No.: 1502
MDL No.: MFCD00149164
Formula:
NaClO₄·H₂O Melting Pt: 130 °C (dec.) Density: 2.02 Storage Temperature: Ambient |
MDL Number:
MFCD00149164 CAS Number: 7791-07-3 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 14,8653 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...