Terephthaloyl dichloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Terephthaloyl chloride
A11224-0B
A11224-30
CAAAAA11224-0BEA
218.04
CAD
CAAAAA11224-0B
CAAAAA11224-30
Terephthaloyl dichloride 99%
Terephthaloyl dichloride
Purity: 99%
CAS No.: 100-20-9
Molecular Formula: C8H4Cl2O2
Formula Weight: 203.02
Flash Point: 180°(356°F)
Melting Point: 79-83°
Boiling Point: 265°
Beilstein Reference: 607796
UN No.: 2928
MDL No.: MFCD00000693
Formula:
C₈H₄Cl₂O₂ MW: 203.02 g/mol Boiling Pt: 266 Melting Pt: 79…83 °C Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000693 CAS Number: 100-20-9 EINECS: 202-829-5 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...