2,3-Dichlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11206-18
A11206-14
A11206-22
CAAAAA11206-18EA
183.28
CAD
CAAAAA11206-18
CAAAA11206-14
CAAAA11206-22
2,3-Dichlorotoluene 98%
2,3-Dichlorotoluene
Purity: 98%
CAS No.: 32768-54-0
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 83°(181°F)
Refractive Index: 1.5510
Melting Point: 4°
Boiling Point: 207-208°
Beilstein Reference: 2412384
UN No.: 3082
MDL No.: MFCD00000546
Formula:
C₇H₆Cl₂ Boiling Pt: 207…208 °C Melting Pt: 4 °C Density: 1.229 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000546 CAS Number: 32768-54-0 EINECS: 251-203-8 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...