Trichloroacetic acid (TCA) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TCA

A11156-0B A11156-0I A11156-30 A11156-36
CAAAAA11156-0BEA 218.04 CAD
CAAAAA11156-0B CAAAAA11156-0I CAAAAA11156-30 CAAAAA11156-36
Trichloroacetic acid (TCA) 99%
Trichloroacetic acid (TCA)

Protein precipitation reagent


Purity: 99% CAS No.: 76-03-9 Molecular Formula: Cl3CCO2H Formula Weight: 163.39 Odor: Acrid pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: 54…58 °C
Density: 1.629
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8 II
Merck Index: 14,9627
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR