Sodium pyruvate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

A11148-18 A11148-30 A11148-36
CAAAAA11148-18EA 121.03 CAD
CAAAAA11148-18 CAAAAA11148-30 CAAAAA11148-36
Sodium pyruvate 99%
Sodium pyruvate

Purity: 99% CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Melting Point: 300° Beilstein Reference: 3568341

Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: >300 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR