Tryptamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11116-09 A11116-18
CAAAA11116-09EA 73 CAD
CAAAA11116-09 CAAAA11116-18
Tryptamine 98+%
Tryptamine

Purity: 98+% CAS No.: 61-54-1 Molecular Formula: C10H12N2 Formula Weight: 160.22 Storage Temperature: 2°C to 8°C Flash Point: 185°(365°F) Melting Point: 111-116° Boiling Point: 137°/0.15mm Beilstein Reference: 125513 Merck Reference: 14,9796 MDL No.: MFCD00005661

Formula: C₁₀H₁₂N₂
Boiling Pt: 137 °C/0.15 mm
Melting Pt: 111…116 °C
Flash Pt: 185 °C(365 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005661
CAS Number: 61-54-1
EINECS: 200-510-5
Merck Index: 14,9796

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR