Trioctylphosphine oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide

A11062-14 A11062-22 A11062-36
CAAAAA11062-14EA 77.1 CAD
CAAAAA11062-14 CAAAAA11062-22 CAAAAA11062-36
Trioctylphosphine oxide 98%
Trioctylphosphine oxide

Purity: 98% CAS No.: 78-50-2 Molecular Formula: C24H51OP Formula Weight: 386.65 Flash Point: 252°(486°F) Odor: Odorless Boiling Point: 212-213°/0.3mm Beilstein Reference: 1796648 MDL No.: MFCD00002083

Formula: C₂₄H₅₁OP
MW: 386.64 g/mol
Boiling Pt: 409
Melting Pt: 52…57 °C
Density: 0.88
Flash Pt: 252 °C (486 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002083
CAS Number: 78-50-2
EINECS: 201-121-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR