2-Nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Nitroaniline

A11055-0B A11055-0I A11055-30
CAAAAA11055-0BEA 132.88 CAD
CAAAAA11055-0B CAAAAA11055-0I CAAAAA11055-30
2-Nitroaniline 98%
2-Nitroaniline

Purity: 98% CAS No.: 88-74-4 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Flash Point: 168°(334°F) Melting Point: 70-74° Boiling Point: 284-286° Beilstein Reference: 509275 Merck Reference: 14,6583 UN No.: 1661 MDL No.: MFCD00007687

Formula: C₆H₆N₂O₂
Boiling Pt: 284…286 °C
Melting Pt: 70…74 °C
Density: 1.255
Flash Pt: 168 °C (334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007687
CAS Number: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1,II
Merck Index: 14,6583

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR