Tryptamine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11030-09
A11030-18
CAAAA11030-09EA
118.34
CAD
CAAAA11030-09
CAAAA11030-18
CAAAA11030-30
Tryptamine hydrochloride 98+%
Tryptamine hydrochloride
Purity: 98+%
CAS No.: 343-94-2
Molecular Formula: C10H12N2.HCl
Formula Weight: 196.68
Melting Point: 252-255°
Beilstein Reference: 3568419
Merck Reference: 14,9796
MDL No.: MFCD00012682
Formula:
C₁₀H₁₃ClN₂ Melting Pt: 252…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012682 CAS Number: 343-94-2 EINECS: 206-446-4 Merck Index: 14,9796 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...