Cyclopentene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11027-AE
A11027-AP
CAAAAA11027-AEEA
84.21
CAD
CAAAAA11027-AE
CAAAAA11027-AP
Cyclopentene 97%
Cyclopentene
Purity: 97%
CAS No.: 142-29-0
Molecular Formula: C5H8
Formula Weight: 68.12
Storage Temperature: 2°C to 8°C
Flash Point: -29°(-20°F)
Refractive Index: 1.4225
Melting Point: -135°
Boiling Point: 44-45°
Beilstein Reference: 635707
UN No.: 2246
MDL No.: MFCD00001394
Formula:
C₅H₈ MW: 68.12 g/mol Boiling Pt: 44…45 °C Melting Pt: –135 °C Density: 0.773 Flash Pt: –29 °C (–20 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001394 CAS Number: 142-29-0 EINECS: 205-532-9 UN: 2246 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...