Barium bromate 97%

Supplier: Thermo Scientific Chemicals
Danger

A11008-22 A11008-36
CAAAAA11008-22EA 97.01 CAD
CAAAAA11008-22 CAAAAA11008-36
Barium bromate 97%
Barium bromate

Purity: 97% CAS No.: 13967-90-3 Molecular Formula: BaBr2O6 Formula Weight: 393.16 Odor: Odorless UN No.: 2719 MDL No.: MFCD00014177

Formula: BaBr₂O₆
Density: 3.99
Storage Temperature: Ambient
MDL Number: MFCD00014177
CAS Number: 13967-90-3
EINECS: 237-750-5
UN: 2719
ADR: 5.1,II
Merck Index: 12,00994

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR