Furfuryl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10968-0B
A10968-18
A10968-30
CAAAAA10968-0BEA
120.4
CAD
CAAAAA10968-0B
CAAAAA10968-18
CAAAAA10968-30
Furfuryl alcohol 98%
Furfuryl alcohol
Purity: 98%
CAS No.: 98-00-0
Molecular Formula: C5H6O2
Formula Weight: 98.10
Flash Point: 65°(149°F)
Refractive Index: 1.4870
Odor: Mild
Melting Point: -31°
Boiling Point: 170-171°
Beilstein Reference: 106291
Merck Reference: 14,4305
UN No.: 2874
MDL No.: MFCD00003252
Formula:
C₅H₆O₂ Boiling Pt: 170…171 °C Melting Pt: –31 °C Density: 1.129 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003252 CAS Number: 98-00-0 EINECS: 202-626-1 UN: 2874 ADR: 6.1,III Merck Index: 14,4305 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...