p-Anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline

A10946-0B A10946-22 A10946-36
CAAAAA10946-0BEA 249.64 CAD
CAAAAA10946-0B CAAAAA10946-22 CAAAAA10946-36
p-Anisidine 99%
p-Anisidine

Purity: 99% CAS No.: 104-94-9 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 122°(251°F) Melting Point: 56-60° Boiling Point: 240-243° Beilstein Reference: 471556 Merck Reference: 14,667 UN No.: 2811 MDL No.: MFCD00007864

Formula: C₇H₉NO
Boiling Pt: 240…243 °C
Melting Pt: 56…60 °C
Density: 1.089
Flash Pt: 122 °C (251 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007864
CAS Number: 104-94-9
EINECS: 203-254-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR