2,4-Dichlorophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2,4-Dichlorophenyl)hydrazine hydrochloride, 2,4-Dichlorophenylhydrazine hydrochloride, 1-(2,4-Dichlorophenyl)hydrazine hydrochloride

A10943-09 A10943-18 A10943-30
CAAAA10943-09EA 119.92 CAD
CAAAA10943-09 CAAAA10943-18 CAAAA10943-30
2,4-Dichlorophenylhydrazinium chloride 98%
2,4-Dichlorophenylhydrazinium chloride

Purity: 98% CAS No.: 5446-18-4 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.49 Melting Point: ca 212° dec. Beilstein Reference: 3708828

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: ca 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012929
CAS Number: 5446-18-4
EINECS: 226-659-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR