2,5-Dibromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10937-09
A10937-18
CAAAAA10937-09EA
85.16
CAD
CAAAAA10937-09
CAAAAA10937-18
2,5-Dibromopyridine 97%
2,5-Dibromopyridine
Purity: 97%
CAS No.: 624-28-2
Molecular Formula: C5H3Br2N
Formula Weight: 236.90
Melting Point: 92-96°
Boiling Point: 249°
MDL No.: MFCD00006221
Formula:
C₅H₃Br₂N MW: 236.89 g/mol Boiling Pt: 249 °C Melting Pt: 92…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006221 CAS Number: 624-28-2 EINECS: 210-839-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...