Diethylenetriaminepentaacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10926-0B A10926-0I A10926-30
CAAAAA10926-0BEA 169.06 CAD
CAAAAA10926-0B CAAAAA10926-0I CAAAAA10926-30
Diethylenetriaminepentaacetic acid 98+%
Diethylenetriaminepentaacetic acid

Purity: 98+% CAS No.: 67-43-6 Molecular Formula: C14H23N3O10 Formula Weight: 393.35 Odor: Odorless Melting Point: ca 220° dec. Beilstein Reference: 1810219 Merck Reference: 14,7125 MDL No.: MFCD00004289

Formula: C₁₄H₂₃N₃O₁₀
Melting Pt: ∼ 220 °C (dec.)
Density: 1.56
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
ADR: 9,III
Merck Index: 14,7125

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR