Potassium hexafluorotitanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium hexafluorotitanate(IV)

A10915-0E A10915-22 A10915-36
CAAAAA10915-0EEA 316 CAD
CAAAAA10915-0E CAAAAA10915-22 CAAAAA10915-36
Potassium hexafluorotitanate 97%
Potassium hexafluorotitanate

Purity: 97% CAS No.: 16919-27-0 Molecular Formula: F6K2Ti Formula Weight: 240.07 Boiling Point: 235-237° UN No.: 3288 MDL No.: MFCD00011380

Formula: F₆K₂Ti
MW: 240.07 g/mol
Boiling Pt: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00011380
CAS Number: 16919-27-0
EINECS: 240-969-9
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR