6-Chloroanthranilic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10890-06
A10890-09
A10890-14
CAAAAA10890-06EA
111.71
CAD
CAAAAA10890-06
CAAAAA10890-09
CAAAAA10890-14
6-Chloroanthranilic acid 97+%
6-Chloroanthranilic acid
Purity: 97+%
CAS No.: 2148-56-3
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Melting Point: 149-152°
Beilstein Reference: 2804041
Formula:
C₇H₆ClNO₂ Boiling Pt: 330 °C (1013 hPa) Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051530 CAS Number: 2148-56-3 EINECS: 218-416-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...