DL-Norleucine 98%
Supplier: Thermo Scientific Chemicals
A10791-14
A10791-22
CAAAAA10791-14EA
104.12
CAD
CAAAAA10791-14
CAAAAA10791-22
CAAAAA10791-36
DL-Norleucine 98%
DL-Norleucine
Purity: 98%
CAS No.: 616-06-8
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Odor: Odorless
Melting Point:
Formula:
C₆H₁₃NO₂ Boiling Pt: 246 °C (1013 hPa) Melting Pt: >300 °C Density: 1.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064422 CAS Number: 616-06-8 EINECS: 210-462-7 Merck Index: 14,6706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...