2,2,2-Trifluoroethanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoro Ethanol, TFE

A10788-0B A10788-18 A10788-22 A10788-36
CAAAAA10788-0BEA 551.42 CAD
CAAAAA10788-0B CAAAAA10788-18 CAAAAA10788-22 CAAAAA10788-36
2,2,2-Trifluoroethanol 99+%
2,2,2-Trifluoroethanol

Purity: 99+% CAS No.: 75-89-8 Molecular Formula: C2H3F3O Formula Weight: 100.04 Flash Point: 29°(84°F) Refractive Index: 1.2900 Melting Point: -44° Boiling Point: 74-75° Beilstein Reference: 1733203 UN No.: 1992 MDL No.: MFCD00004672

Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 74…75 °C
Melting Pt: –44 °C
Density: 1.391
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 1992
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR