Methyl-5-chloro-2-methoxybenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Chloro-2-methoxybenzoic acid methyl ester, Methyl 5-chloro-2-methoxybenzoate

A10787-14 A10787-22 A10787-36
CAAAA10787-14EA 67.78 CAD
CAAAA10787-14 CAAAA10787-22 CAAAA10787-36
Methyl-5-chloro-2-methoxybenzoate 98%
Methyl 5-chloro-2-methoxybenzoate

Purity: 98% CAS No.: 33924-48-0 Molecular Formula: C9H9ClO3 Formula Weight: 200.62 Flash Point: 110°(230°F) Refractive Index: 1.5460 Boiling Point: 235-240° Beilstein Reference: 2719088 MDL No.: MFCD00017511

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Boiling Pt: 235…240 °C
Density: 1.259
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017511
CAS Number: 33924-48-0
EINECS: 251-743-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR