Manganese(IV) oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

A10765-0E A10765-36 A10765-A9
CAAAAA10765-0EEA 297.04 CAD
CAAAAA10765-0E CAAAAA10765-22 CAAAAA10765-36 CAAAAA10765-A9
Manganese(IV) oxide 98%
Manganese (IV) oxide

Purity: 98% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Melting Point: 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463

Formula: MnO₂
Melting Pt: 535 °C (dec.)
Density: 5.026
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR