(±)-2-Bromoisovaleric acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-3-methylbutyricacid, 2-Bromo-3-methylbutyric acid
A10756-09
A10756-18
CAAAA10756-09EA
59.57
CAD
CAAAA10756-09
CAAAA10756-18
(±)-2-Bromoisovaleric acid 96%
(±)-2-Bromoisovaleric acid
Purity: 96%
CAS No.: 565-74-2
Molecular Formula: C5H9BrO2
Formula Weight: 181.04
Odor: Acrid
Melting Point: 36-42°
Boiling Point: 124-126°/20mm
Merck Reference: 14,1422
UN No.: 3261
MDL No.: MFCD00004210
Formula:
C₅H₉BrO₂ Boiling Pt: 124…126 °C/20 mm Melting Pt: 36…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004210 CAS Number: 565-74-2 EINECS: 209-291-0 UN: 3261 ADR: "8",III Merck Index: 14,1422 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...