2,3-Dichlorophenylhydrazinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10717-06
A10717-14
A10717-22
CAAAA10717-06EA
105.39
CAD
CAAAA10717-06
CAAAA10717-14
CAAAA10717-22
2,3-Dichlorophenylhydrazinium chloride 97%
2,3-Dichlorophenylhydrazinium chloride
Purity: 97%
CAS No.: 21938-47-6
Molecular Formula: C6H6Cl2N2.HCl
Formula Weight: 213.49
Melting Point: ca 204° dec.
Beilstein Reference: 3708910
Formula:
C₆H₇Cl₃N₂ Melting Pt: ∼ 204 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052265 CAS Number: 21938-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...