D-(+)-Camphor 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10708-14
A10708-22
A10708-36
CAAAAA10708-14EA
42.19
CAD
CAAAAA10708-14
CAAAAA10708-22
CAAAAA10708-36
D-(+)-Camphor 98%
D(+)-Camphor
Purity: 98%
CAS No.: 464-49-3
Molecular Formula: C10H16O
Formula Weight: 152.24
Flash Point: 65°(149°F)
Odor: Camphor-like
Boiling Point: 204°
Beilstein Reference: 2042745
Merck Reference: 14,1732
Optical Rotation: +43° (c=10 in ethanol)
UN No.: 2717
MDL No.: MFCD00064149
Formula:
C₁₀H₁₆O Boiling Pt: 204 °C Melting Pt: 175…180 °C Density: 0.990 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064149 CAS Number: 464-49-3 EINECS: 207-355-2 UN: 2717 ADR: 4.1,III Merck Index: 14,1732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...