Ammonium thiocyanate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

A10632-0B A10632-0I A10632-30
CAAAAA10632-0BEA 271.76 CAD
CAAAAA10632-0B CAAAAA10632-0I CAAAAA10632-30
Ammonium thiocyanate 98+%
Ammonium thiocyanate

Purity: 98+% CAS No.: 1762-95-4 Molecular Formula: CH4N2S Formula Weight: 76.12 Odor: Odorless Melting Point: 147-149° Beilstein Reference: 3595135 Merck Reference: 14,561 MDL No.: MFCD00011428

Formula: NH₄SCN
Melting Pt: 147…149 °C
Density: 1.305
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 14,561

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR