Ammonium cobalt (II) sulfate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: di-Ammonium cobalt (II) sulfate hexahydrate, Ammonium cobaltous sulphate hexahydrate, Ammonium cobaltous sulfate hexahydrate, di-Ammonium cobalt (II) sulphate hexahydrate

A10627-14 A10627-22 A10627-36
CAAAAA10627-14EA 80.58 CAD
CAAAAA10627-14 CAAAAA10627-22 CAAAAA10627-36
Ammonium cobalt (II) sulfate hexahydrate 98%
Ammonium cobalt (II) sulphate hexahydrate

Purity: 98% CAS No.: 13586-38-4 Molecular Formula: (NH4)2Co(SO4)2.6H2O Formula Weight: 395.21 (287.13anhy) Odor: Odorless Melting Point: ca 120° dec. UN No.: 3077 MDL No.: MFCD00151049

Formula: H₈CoN₂O₈S₂·6H₂O
Melting Pt: ca 120 °C (dec.)
Density: 1.902
Storage Temperature: Ambient
MDL Number: MFCD00151049
CAS Number: 13586-38-4
EINECS: 237-043-1
UN: 3077
ADR: "9",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR