DL-Threonine 99%

Supplier: Thermo Scientific Chemicals

A10606-14 A10606-22 A10606-36
CAAAA10606-14EA 68.41 CAD
CAAAA10606-14 CAAAA10606-22 CAAAA10606-36
DL-Threonine 99%
DL-Threonine

Purity: 99% CAS No.: 80-68-2 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: ca 244° dec. Beilstein Reference: 1721647 Merck Reference: 14,9380 MDL No.: MFCD00063722

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: ∼ 244 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063722
CAS Number: 80-68-2
EINECS: 201-300-6
Merck Index: 14,9380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR