Methyl salicylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10578-0E
A10578-30
A10578-36
CAAAAA10578-0EEA
254.38
CAD
CAAAAA10578-0E
CAAAAA10578-30
CAAAAA10578-36
Methyl salicylate 98%
Methyl salicylate
Purity: 98%
CAS No.: 119-36-8
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point: 99°(210°F)
Refractive Index: 1.5360
Odor: Characteristic
Melting Point: -9°
Boiling Point: 224°
Beilstein Reference: 971516
Merck Reference: 14,6120
MDL No.: MFCD00002214
Formula:
2-(HO)C₆H₄CO₂CH₃ Boiling Pt: 224 °C Melting Pt: –9 °C Density: 1.180 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002214 CAS Number: 119-36-8 EINECS: 204-317-7 Merck Index: 14,6120 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...