Sodium bromide, (max. 1% H₂O) 99+% (dry weight)

Supplier: Thermo Scientific Chemicals

A10552-0B A10552-0I A10552-30
CAAAAA10552-0BEA 148.99 CAD
CAAAAA10552-0B CAAAAA10552-0I CAAAAA10552-30
Sodium bromide, (max. 1% H₂O) 99+% (dry weight)
Sodium bromide

Purity: 99+% CAS No.: 7647-15-6 Molecular Formula: NaBr Formula Weight: 102.89 Refractive Index: 1.6412 Melting Point: 755° Boiling Point: 1390° Merck Reference: 14,8594 MDL No.: MFCD00003475

Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396
Melting Pt: 753
Density: 3.203
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 14,8594

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR