Methyl-4-iodo-2-methoxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate

A10549-06 A10549-14
CAAAAA10549-22EA 959.46 CAD
CAAAAA10549-22 CAAAA10549-06 CAAAA10549-14
Methyl-4-iodo-2-methoxybenzoate 98%
Methyl 4-iodo-2-methoxybenzoate

Purity: 98% CAS No.: 35387-93-0 Molecular Formula: C9H9IO3 Formula Weight: 292.07 Melting Point: 93-95° Beilstein Reference: 2692711

Formula: C₉H₉IO₃
MW: 292.07 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00052925
CAS Number: 35387-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR