Ammonium persulfate (APS) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Peroxidisulfuric acid diammonium salt, Ammonium persulfate, Peroxidisulphuric acid diammonium salt, Ammonium persulphate
A10533-0B
A10533-0I
A10533-36
CAAAAA10533-0BEA
131.46
CAD
CAAAAA10533-0B
CAAAAA10533-0I
CAAAAA10533-36
Ammonium persulfate (APS) 98%
Ammonium peroxodisulphate (APS)
Purity: 98%
CAS No.: 7727-54-0
Molecular Formula: H8N2O8S2
Formula Weight: 228.19
Flash Point: None
Refractive Index: 1.50
Odor: Odorless
Melting Point: 120° dec.
Merck Reference: 14,541
UN No.: 1444
MDL No.: MFCD00003390
Formula:
(NH₄)₂S₂O₈ MW: 228.2 g/mol Melting Pt: 120 °C (dec.) Density: 1.982 Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1 III Merck Index: 14,541 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...