2,6-Lutidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10478-AC
A10478-AE
A10478-AP
CAAAAA10478-ACEA
40.45
CAD
CAAAAA10478-AC
CAAAAA10478-AE
CAAAAA10478-AP
2,6-Lutidine 98+%
2,6-Lutidine
Purity: 98+%
CAS No.: 108-48-5
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 33°(91°F)
Refractive Index: 1.4970
Odor: Unpleasant
Melting Point: -7°
Boiling Point: 143-145°
Beilstein Reference: 105690
Merck Reference: 14,5616
UN No.: 1993
MDL No.: MFCD00006345
Formula:
C₇H₉N Boiling Pt: 143…145 °C Melting Pt: –7 °C Density: 0.923 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006345 CAS Number: 108-48-5 EINECS: 203-587-3 UN: 1993 ADR: 3,III Merck Index: 14,5616 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...