(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10459-09
A10459-18
A10459-30
CAAAAA10459-09EA
106.97
CAD
CAAAAA10459-09
CAAAAA10459-18
CAAAAA10459-30
(-)-Quinine, anhydrous 99% (total base) may cont. up to 5% dihydroquinine
(-)-Quinine
A potassium channel blocker
Purity: 99%
CAS No.: 130-95-0
Molecular Formula: C20H24N2O2
Formula Weight: 324.42
Melting Point: ca 177° dec.
Beilstein Reference: 91867
Merck Reference: 14,8061
Optical Rotation: -166° (c=1 in ethanol)
MDL No.: MFCD00198096
Formula:
C₂₀H₂₄N₂O₂ Melting Pt: ca 177 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00198096 CAS Number: 130-95-0 EINECS: 205-003-2 Merck Index: 14,8061 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...