Lithium sulfate monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A10410-0E A10410-22 A10410-36
CAAAAA10410-0EEA 729.96 CAD
CAAAAA10410-0E CAAAAA10410-22 CAAAAA10410-36
Lithium sulfate monohydrate 99%
Lithium sulphate monohydrate

Purity: 99% CAS No.: 10102-25-7 Molecular Formula: Li2SO4.H2O Formula Weight: 127.96 (109.94anhy) Odor: Odorless Melting Point: 130° -H2O MDL No.: MFCD00149766

Formula: Li₂SO₄·H₂O
Melting Pt: 130 °C (–H₂O of crystall.)
Density: 2.06
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4
Merck Index: 14,5541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR