(±)-3-Bromocyclohexene 95% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromocyclohexene

Stabilization: Stabilised with propylene oxide 300-1000 ppm

A10370-06 A10370-14 A10370-22
CAAAAA10370-06EA 78.05 CAD
CAAAAA10370-06 CAAAAA10370-14 CAAAAA10370-22
(±)-3-Bromocyclohexene 95% stabilised
(±)-3-Bromocyclohexene

Purity: 95% CAS No.: 1521-51-3 Molecular Formula: C6H9Br Formula Weight: 161.05 Storage Temperature: 2°C to 8°C Flash Point: 54°(129°F) Refractive Index: 1.5310 Boiling Point: 64-65°/15mm Beilstein Reference: 635953 UN No.: 1993 MDL No.: MFCD00013775

Formula: C₆H₉Br
Boiling Pt: 64…65 °C/15 mm
Density: 1.400
Flash Pt: 54 °C (129 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013775
CAS Number: 1521-51-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR