2,4-Dichloro-6-methylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10363-14
A10363-22
CAAAA10363-14EA
143.62
CAD
CAAAA10363-14
CAAAA10363-22
2,4-Dichloro-6-methylpyrimidine 98%
2,4-Dichloro-6-methylpyrimidine
Purity: 98%
CAS No.: 5424-21-5
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Flash Point:
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Boiling Pt: 219 °C Melting Pt: 44…47 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006064 CAS Number: 5424-21-5 EINECS: 226-563-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...