2-(4-Hydroxyphenylazo)benzoic acid 98+%
Supplier: Thermo Scientific Chemicals
A10333-09
A10333-18
CAAAA10333-09EA
86.58
CAD
CAAAA10333-09
CAAAA10333-18
2-(4-Hydroxyphenylazo)benzoic acid 98+%
2-(4-Hydroxyphenylazo)benzoic acid
Indicator used in determination of serum albumin
Purity: 98+%
CAS No.: 1634-82-8
Molecular Formula: C13H10N2O3
Formula Weight: 242.23
Melting Point: 205-207°
Beilstein Reference: 1842696
Formula:
C₁₃H₁₀N₂O₃ Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002428 CAS Number: 1634-82-8 EINECS: 216-655-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...