Magnesium nitrate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A10329-0B A10329-0I A10329-30
CAAAAA10329-0BEA 107.91 CAD
CAAAAA10329-0B CAAAAA10329-0I CAAAAA10329-30
Magnesium nitrate hexahydrate 98%
Magnesium nitrate hexahydrate

Purity: 98% CAS No.: 13446-18-9 Molecular Formula: Mg(NO3)2.6H2O Formula Weight: 256.41 (148.32anhy) Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00149779

Formula: Mg(NO₃)₂·6H₂O
Boiling Pt: 330 °C (dec.)
Melting Pt: 89 °C
Density: 1.46
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
ADR: 5.1,III
Merck Index: 14,5674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR