Propargyl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Propyn-1-ol, Prop-2-yn-1-ol
A10295-AE
A10295-AP
CAAAAA10295-AEEA
39.5
CAD
CAAAAA10295-AE
CAAAAA10295-AP
Propargyl alcohol 99%
Propargyl alcohol
Purity: 99%
CAS No.: 107-19-7
Molecular Formula: C3H4O
Formula Weight: 56.06
Storage Temperature: 2°C to 8°C
Flash Point: 33°(91°F)
Refractive Index: 1.4320
Melting Point: -53°
Boiling Point: 114-115°
Beilstein Reference: 506003
Merck Reference: 14,7809
UN No.: 2929
MDL No.: MFCD00002912
Formula:
C₃H₄O MW: 56.06 g/mol Boiling Pt: 114…115 °C Melting Pt: –53 °C Density: 0.949 Flash Pt: 33 °C (91 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002912 CAS Number: 107-19-7 EINECS: 203-471-2 UN: 2929 ADR: 6.1,II Merck Index: 14,7809 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...